The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036007.2 | NP_001031084.1 | 1 - 2 | 717 | 97 |
NM_001332604.1 | NP_001320605.1 | 2 - 3 | 842 | 97 | |
NM_102197.5 | NP_173762.4 | 2 - 3 | 842 | 97 | |
NM_180621.3 | NP_850952.1 | 3 - 4 | 744 | 97 | |
GenBank mRNA | AY099769.1 | - | 2 - 3 | 497 | 97 |
BT000203.1 | - | 2 - 3 | 397 | 97 | |
KJ138989.1 | - | 2 - 3 | 397 | 97 |