Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATPI4K ALPHA
Entrez Gene ID
841357
Gene Name
Phosphatidylinositol 3- and 4-kinase family protein
Gene Aliases
AT1G49340
Chromosome Location
Chr.1: 18252050 - 18264672 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333375.1 NP_001323160.1 1 - 2 1304 62
NM_001333376.1 NP_001323161.1 1 - 2 1290 62
NM_103824.4 NP_175359.2 1 - 2 1302 62
NM_180629.2 NP_850960.1 1 - 2 1304 62

View Full Product Details