Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAL3
Entrez Gene ID
830302
Gene Name
phenyl alanine ammonia-lyase 3
Gene Aliases
AT5G04230, ATPAL3, F21E1.150, F21E1_150, phenyl alanine ammonia-lyase 3
Chromosome Location
Chr.5: 1160569 - 1163957 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203294.1 NP_001190223.1 1 - 2 563 70
NM_120505.4 NP_196043.2 1 - 2 628 70
GenBank mRNA AY528562.1 - 1 - 2 366 70

View Full Product Details