Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WCRKC2
Entrez Gene ID
830305
Gene Name
WCRKC thioredoxin 2
Gene Aliases
AT5G04260, T19N18.5, WCRKC thioredoxin 2
Chromosome Location
Chr.5: 1178644 - 1180060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120508.4 NP_196046.2 3 - 4 490 71
GenBank mRNA AY063850.1 - 3 - 4 466 71
AY117290.1 - 3 - 4 452 71

View Full Product Details