Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KTF1
Entrez Gene ID
830308
Gene Name
kow domain-containing transcription factor 1
Gene Aliases
AT5G04290, SPT5-LIKE, SPT5L, T19N18.20, T19N18_20, kow domain-containing transcription factor 1
Chromosome Location
Chr.5: 1195969 - 1202936 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342760.1 NP_001332032.1 8 - 9 1487 70
NM_120511.1 NP_196049.1 8 - 9 1387 70

View Full Product Details