The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001342767.1 | NP_001330089.1 | 19 - 20 | 2174 | 77 |
NM_120518.5 | NP_196056.2 | 19 - 20 | 2187 | 77 |