Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAMT1
Entrez Gene ID
830316
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT5G04370, T19N18.100, T19N18_100
Chromosome Location
Chr.5: 1231609 - 1234147 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036754.2 NP_001031831.1 4 - 5 1042 160
NM_001342768.1 NP_001331549.1 3 - 4 1354 160
NM_120519.3 NP_196057.1 4 - 5 1244 160
GenBank mRNA AK230364.1 - 4 - 5 1042 160

View Full Product Details