Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DME
Entrez Gene ID
830335
Gene Name
HhH-GPD base excision DNA repair family protein
Gene Aliases
AT5G04560, AT5G04570, AT5G04580, DEMETER, T32M21.160, T32M21_160
Chromosome Location
Chr.5: 1309099 - 1318520 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085058.2 NP_001078527.1 18 - 19 6111 110
NM_001342781.1 NP_001331593.1 18 - 19 6116 110
NM_120538.2 NP_196076.2 16 - 17 6526 110
GenBank mRNA DQ335243.1 - 18 - 19 6023 110

View Full Product Details