Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIOF
Entrez Gene ID
830339
Gene Name
biotin F
Gene Aliases
7-KETO-8-AMINOPELARGONIC ACID (KAPA) SYNTHASE, AT5G04620, ATBIOF, BIO4, T1E3.3, biotin 4, biotin F
Chromosome Location
Chr.5: 1327093 - 1330151 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342782.1 NP_001332170.1 4 - 5 909 107
NM_120544.3 NP_196082.2 3 - 4 1142 107
NM_203002.4 NP_974731.1 4 - 5 931 107
GenBank mRNA AK119095.1 - 3 - 4 1142 107
BT026507.1 - 2 - 3 333 107
DQ017966.1 - 4 - 5 750 107

View Full Product Details