Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAD8
Entrez Gene ID
830441
Gene Name
fatty acid desaturase 8
Gene Aliases
AT5G05580, AtFAD8, MOP10.12, MOP10_12, OMEGA-3 FATTY ACID DESATURASE, fatty acid desaturase 8
Chromosome Location
Chr.5: 1664040 - 1666531 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203301.2 NP_001190230.1 3 - 4 914 113
NM_120640.4 NP_196177.1 3 - 4 892 113
GenBank mRNA AF361837.1 - 3 - 4 807 113
AY078043.1 - 3 - 4 623 113
D17578.1 - 3 - 4 760 113
L27158.1 - 3 - 4 683 113

View Full Product Details