Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBE
Entrez Gene ID
830494
Gene Name
C2H2 and C2HC zinc fingers superfamily protein
Gene Aliases
AT5G06070, RAB, RABBIT EARS
Chromosome Location
Chr.5: 1828150 - 1829186 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120689.2 NP_568161.1 1 - 1 737 159
GenBank mRNA AB107371.1 - 1 - 1 737 159

View Full Product Details