Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AtPOT1b
Entrez Gene ID
830519
Gene Name
Nucleic acid-binding, OB-fold-like protein
Gene Aliases
AT5G06310, MHF15.17, MHF15_17, protection of telomeres 1b
Chromosome Location
Chr.5: 1928343 - 1930951 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120714.4 NP_196249.2 7 - 8 1075 67
GenBank mRNA AK221379.1 - 7 - 8 1027 67
AY884594.1 - 7 - 8 950 67
BT012568.1 - 7 - 8 950 67

View Full Product Details