Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLA17
Entrez Gene ID
830527
Gene Name
FASCICLIN-like arabinogalactan protein 17 precursor
Gene Aliases
AT5G06390, FASCICLIN-like arabinogalactan protein 17 precursor, MHF15.9, MHF15_9
Chromosome Location
Chr.5: 1952505 - 1955796 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120722.4 NP_196257.2 2 - 3 1857 91
GenBank mRNA BT015342.1 - 2 - 3 1465 91

View Full Product Details