Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBA 2
- Entrez Gene ID
-
830534
- Gene Name
- ubiquitin activating enzyme 2
- Gene Aliases
-
ARABIDOPSIS THALIANA UBIQUITIN ACTIVATING ENZYME 2, ARABIDOPSIS THALIANA UBIQUITIN ACTIVATING ENZYME 2, AT5G06460, ATUBA2, MHF15.2, MHF15_2, ubiquitin activating enzyme 2
- Chromosome Location
-
Chr.5: 1969960 - 1974527 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120729.4 |
NP_568168.1 |
3 - 4 |
668 |
110 |
View Full Product Details