The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001342930.1 | NP_001331523.1 | 5 - 6 | 391 | 109 |
NM_120800.4 | NP_196335.2 | 7 - 8 | 946 | 109 | |
GenBank mRNA | AY244746.1 | - | 7 - 8 | 776 | 109 |
FJ708772.1 | - | 7 - 8 | 591 | 109 |