Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR21
Entrez Gene ID
830612
Gene Name
response regulator 21
Gene Aliases
ARR21, AT5G07210, T28J14.150, T28J14_150, response regulator 21
Chromosome Location
Chr.5: 2252237 - 2256018 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342931.1 NP_001331229.1 5 - 6 1542 88
NM_120803.1 NP_196338.1 5 - 6 1566 88

View Full Product Details