The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_120804.3 | NP_196339.1 | 1 - 2 | 485 | 80 |
GenBank mRNA | AF428411.1 | - | 1 - 2 | 387 | 80 |