Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ML4
Entrez Gene ID
830620
Gene Name
MEI2-like 4
Gene Aliases
AML4, AT5G07290, MEI2-like 4, T28J14.230, T28J14_230
Chromosome Location
Chr.5: 2293114 - 2298976 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120811.5 NP_196346.1 5 - 6 1580 68
GenBank mRNA AK226960.1 - 5 - 6 1324 68

View Full Product Details