Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCT2
Entrez Gene ID
830687
Gene Name
MEI2 C-terminal RRM only like 2
Gene Aliases
AT5G07930, F13G24.130, MEI2 C-terminal RRM only like 2
Chromosome Location
Chr.5: 2530834 - 2532742 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085075.1 NP_001078544.1 2 - 3 557 135
NM_001203324.1 NP_001190253.1 2 - 3 588 135
NM_120875.1 NP_196410.1 2 - 3 557 135
GenBank mRNA DQ446931.1 - 2 - 3 557 135

View Full Product Details