Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFH3
Entrez Gene ID
816693
Gene Name
SEC14-like 3
Gene Aliases
AT2G21540, ATSFH3, F3K23.30, SEC14-LIKE 3, SEC14-like 3
Chromosome Location
Chr.2: 9220505 - 9224496 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036312.2 NP_001031389.1 11 - 12 1506 66
NM_001202642.2 NP_001189571.1 10 - 11 1488 66
NM_001335761.1 NP_001318263.1 11 - 12 1373 66
NM_001335762.1 NP_001323476.1 10 - 11 1369 66
NM_001335763.1 NP_001323475.1 11 - 12 1302 66
NM_127726.2 NP_565514.1 11 - 12 1387 66
GenBank mRNA AK176420.1 - 11 - 12 1353 66
AY057587.1 - 11 - 12 1389 66

View Full Product Details