Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EME1A
Entrez Gene ID
816716
Gene Name
essential meiotic endonuclease 1A
Gene Aliases
AT2G21800, ATEME1A, F7D8.12, F7D8_12, essential meiotic endonuclease 1A
Chromosome Location
Chr.2: 9298078 - 9301926 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202643.1 NP_001189572.1 3 - 4 496 74
NM_001335775.1 NP_001323848.1 3 - 4 561 74
NM_127749.5 NP_179771.2 3 - 4 573 74
GenBank mRNA AK228007.1 - 3 - 4 496 74
BT006456.1 - 3 - 4 455 74
FJ936556.1 - 3 - 4 455 74

View Full Product Details