Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACS4
Entrez Gene ID
816812
Gene Name
1-aminocyclopropane-1-carboxylate synthase 4
Gene Aliases
1-AMINOCYCLOPROPANE-1-CARBOXYLATE SYNTHASE, 1-AMINOCYCLOPROPANE-1-CARBOXYLIC ACID SYNTHASE POLYPEPTIDE, 1-AMINOCYCLOPROPANE-1-CARBOXYLIC ACID SYNTHASE POLYPEPTIDE, 1-aminocyclopropane-1-carboxylate synthase 4, ACC4, AT2G22810, ATACS4, T30L20.7
Chromosome Location
Chr.2: 9717602 - 9719594 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127846.2 NP_179866.1 1 - 2 269 68
GenBank mRNA AF332404.1 - 1 - 2 147 68
U23481.1 - 1 - 2 147 68

View Full Product Details