The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_127849.4 | NP_179869.1 | 2 - 3 | 902 | 114 |
GenBank mRNA | AB493560.1 | - | 2 - 3 | 541 | 114 |
AY102634.1 | - | 2 - 3 | 613 | 114 |