Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL11
Entrez Gene ID
816828
Gene Name
serine carboxypeptidase-like 11
Gene Aliases
AT2G22970, T20K9.19, serine carboxypeptidase-like 11
Chromosome Location
Chr.2: 9774749 - 9778561 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036324.1 NP_001031401.1 9 - 10 882 125
NM_001084476.1 NP_001077945.1 9 - 10 882 125
NM_001335831.1 NP_001323542.1 9 - 10 887 125
NM_127862.4 NP_179880.1 9 - 10 986 125
GenBank mRNA AY039596.1 - 9 - 10 875 125
AY113074.1 - 9 - 10 860 125
BT000747.1 - 9 - 10 882 125

View Full Product Details