Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNG1
Entrez Gene ID
816830
Gene Name
sinapoylglucose 1
Gene Aliases
AT2G22990, MALATE SINAPOYLTRANSFERASE, SCPL8, SERINE CARBOXYPEPTIDASE-LIKE 8, T20K9.18, sinapoylglucose 1
Chromosome Location
Chr.2: 9786087 - 9790335 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161057.2 NP_001154529.1 8 - 9 1239 82
NM_127864.3 NP_565546.3 8 - 9 885 82
NM_179703.2 NP_850034.1 9 - 10 1135 82
NM_179704.1 NP_850035.1 9 - 10 939 82
NM_179705.1 NP_850036.1 9 - 10 939 82
NM_201787.2 NP_973516.2 9 - 10 912 82
GenBank mRNA AF275313.1 - 9 - 10 925 82
AY035052.1 - 9 - 10 939 82
AY051060.1 - 9 - 10 862 82

View Full Product Details