Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl10
Entrez Gene ID
816831
Gene Name
serine carboxypeptidase-like 10
Gene Aliases
AT2G23000, F21P24.6, F21P24_6, serine carboxypeptidase-like 10
Chromosome Location
Chr.2: 9792166 - 9796408 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335834.1 NP_001324624.1 7 - 8 836 144
NM_127865.3 NP_179883.1 7 - 8 790 144

View Full Product Details