Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP17
Entrez Gene ID
816845
Gene Name
arabinogalactan protein 17
Gene Aliases
ARABINOGALACTAN PROTEIN 17, AT2G23130, ATAGP17, T20D16.24, arabinogalactan protein 17
Chromosome Location
Chr.2: 9844134 - 9845385 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127877.3 NP_179894.1 1 - 2 748 88
GenBank mRNA AF305939.1 - 1 - 2 471 88
AY086875.1 - 1 - 2 532 88
BT002926.1 - 1 - 2 532 88
BT008601.1 - 1 - 2 471 88

View Full Product Details