Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NRAMP3
- Entrez Gene ID
-
816847
- Gene Name
- natural resistance-associated macrophage protein 3
- Gene Aliases
-
AT2G23150, ATNRAMP3, METAL TRANSPORTER NRAMP3, T20D16.22, T20D16_22, natural resistance-associated macrophage protein 3
- Chromosome Location
-
Chr.2: 9856149 - 9858778 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127879.4 |
NP_179896.1 |
3 - 4 |
1542 |
60 |
GenBank mRNA |
AY058203.1 |
- |
3 - 4 |
1338 |
60 |
View Full Product Details