Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CLF
- Entrez Gene ID
-
816870
- Gene Name
- SET domain-containing protein
- Gene Aliases
-
AT2G23380, CURLY LEAF, F26B6.3, F26B6_3, ICU1, INCURVATA 1, SDG1, SET1, SETDOMAIN 1, SETDOMAIN GROUP 1
- Chromosome Location
-
Chr.2: 9955092 - 9960386 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127902.6 |
NP_179919.1 |
5 - 6 |
1183 |
98 |
View Full Product Details