Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAPRT2
Entrez Gene ID
816874
Gene Name
nicotinate phosphoribosyltransferase 2
Gene Aliases
AT2G23420, F26B6.7, F26B6_7, nicotinate phosphoribosyltransferase 2
Chromosome Location
Chr.2: 9971603 - 9975207 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335851.1 NP_001323701.1 13 - 14 1697 100
NM_127906.6 NP_179923.2 13 - 14 1698 100
GenBank mRNA BT001932.1 - 13 - 14 1482 100

View Full Product Details