Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RS2Z32
Entrez Gene ID
824518
Gene Name
RNA-binding (RRM/RBD/RNP motifs) family protein with retrovirus zinc finger-like domain-containing protein
Gene Aliases
AT3G53500, At-RS2Z, RSZ32, arginine/serine-rich zinc knuckle-containing protein 32
Chromosome Location
Chr.3: 19834240 - 19837088 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339619.1 NP_001326575.1 5 - 6 1188 75
NM_115210.6 NP_190918.3 6 - 7 700 75
NM_180684.4 NP_851015.1 6 - 7 912 75
GenBank mRNA AK316697.1 - 6 - 7 624 75
AY095996.1 - 6 - 7 841 75
BT001034.1 - 4 - 5 414 75

View Full Product Details