Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPT9
Entrez Gene ID
832126
Gene Name
isopentenyltransferase 9
Gene Aliases
ARABIDOPSIS THALIANA ISOPENTENYLTRANSFERASE 9, AT5G20040, ATIPT9, F28I16.190, F28I16_190, isopentenyltransferase 9
Chromosome Location
Chr.5: 6767701 - 6771190 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203415.1 NP_001190344.1 9 - 10 1379 87
NM_122011.4 NP_568390.2 9 - 10 1341 87
NM_180712.3 NP_851043.1 9 - 10 1341 87
GenBank mRNA AB062615.1 - 9 - 10 1151 87
AF360208.1 - 9 - 10 1266 87
AY040056.1 - 9 - 10 1151 87

View Full Product Details