Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL34
Entrez Gene ID
832385
Gene Name
serine carboxypeptidase-like 34
Gene Aliases
AT5G23210, MKD15.7, MKD15_7, serine carboxypeptidase-like 34
Chromosome Location
Chr.5: 7810645 - 7815089 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085146.2 NP_001078615.1 6 - 7 1246 80
NM_001085147.2 NP_001078616.1 6 - 7 747 80
NM_001343793.1 NP_001331558.1 5 - 6 1360 80
NM_122227.2 NP_197712.2 6 - 7 747 80
NM_180731.3 NP_851062.2 6 - 7 1267 80
GenBank mRNA AK228998.1 - 6 - 7 747 80

View Full Product Details