Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYSD2
Entrez Gene ID
832872
Gene Name
cysteine synthase D2
Gene Aliases
AT5G28020, ATCYSD2, CYSTEINE SYNTHASE, CYSTEINE SYNTHASE D2, F15F15.90, F15F15_90, cysteine synthase D2
Chromosome Location
Chr.5: 10026152 - 10028722 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036879.2 NP_001031956.1 9 - 10 942 142
NM_001036880.3 NP_001031957.1 9 - 10 1025 142
NM_001036881.1 NP_001031958.1 6 - 7 762 142
NM_001085159.1 NP_001078628.1 9 - 10 919 142
NM_122685.3 NP_198154.1 9 - 10 1068 142
NM_180756.3 NP_851087.1 8 - 9 1252 142
GenBank mRNA AB024283.1 - 8 - 9 1241 142
AF424598.1 - 9 - 10 926 142

View Full Product Details