Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHT1
Entrez Gene ID
834078
Gene Name
lysine histidine transporter 1
Gene Aliases
AT5G40780, K1B16.3, K1B16_3, LYSINE AND HISTIDINE SPECIFIC TRANSPORTER, lysine histidine transporter 1
Chromosome Location
Chr.5: 16323567 - 16327382 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344353.1 NP_001318716.1 2 - 3 560 63
NM_001344354.1 NP_001332682.1 2 - 3 372 63
NM_180778.4 NP_851109.1 2 - 3 456 63
GenBank mRNA AF367281.1 - 2 - 3 303 63
AY133550.1 - 2 - 3 200 63

View Full Product Details