Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAT5
Entrez Gene ID
834946
Gene Name
peroxisomal 3-keto-acyl-CoA thiolase 2
Gene Aliases
3-KETO-ACYL-COA THIOLASE, 3-KETO-ACYL-COENZYME A THIOLASE 5, AT5G48880, K24G6.22, K24G6_22, PEROXISOMAL 3-KETO-ACYL-COA THIOLASE 2, PEROXISOMAL-3-KETO-ACYL-COA THIOLASE 1, PKT1, PKT2, peroxisomal 3-keto-acyl-CoA thiolase 2
Chromosome Location
Chr.5: 19814395 - 19817955 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036960.2 NP_001032037.1 8 - 9 723 58
NM_001344808.1 NP_001331840.1 8 - 9 895 58
NM_124265.5 NP_568704.2 7 - 8 1590 58
NM_180826.4 NP_851157.1 7 - 8 740 58
GenBank mRNA AF062590.1 - 7 - 8 620 58
AF062591.1 - 7 - 8 839 58
AY079026.1 - 7 - 8 725 58
AY136455.1 - 7 - 8 628 58
BT002594.1 - 6 - 7 527 58

View Full Product Details