Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMF2
Entrez Gene ID
835198
Gene Name
VEFS-Box of polycomb protein
Gene Aliases
AT5G51230, AT5G51240, AtEMF2, CYR1, CYTOKININ RESISTANT 1, EMBRYONIC FLOWER 2, MWD22.18, MWD22_18, VEF2
Chromosome Location
Chr.5: 20823644 - 20829644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125941.1 NP_001119413.1 20 - 21 2101 73
NM_124502.3 NP_199936.2 21 - 22 2093 73
NM_180837.3 NP_851168.1 21 - 22 2200 73
GenBank mRNA AB053171.1 - 21 - 22 2045 73
AY140086.1 - 21 - 22 2042 73
BT008388.1 - 20 - 21 1782 73

View Full Product Details