Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT5G51630
Entrez Gene ID
835237
Gene Name
Disease resistance protein (TIR-NBS-LRR class) family
Gene Aliases
AT5G51630, K17N15.18, K17N15_18
Chromosome Location
Chr.5: 20969128 - 20975940 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203587.1 NP_001190516.1 6 - 7 2514 131
NM_001344946.1 NP_001330479.1 5 - 6 2895 131
NM_001344947.1 NP_001330480.1 5 - 6 2701 131
NM_001344948.1 NP_001330481.1 5 - 6 2636 131
NM_124542.3 NP_199976.2 5 - 6 2840 131
NM_180841.3 NP_851172.1 4 - 5 3230 131
GenBank mRNA AK228948.1 - 4 - 5 2383 131

View Full Product Details