Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIP2
Entrez Gene ID
835591
Gene Name
potassium channel tetramerization domain-containing protein / pentapeptide repeat-containing protein
Gene Aliases
AT5G55000, MBG8.27, MBG8_27
Chromosome Location
Chr.5: 22318305 - 22321805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124882.4 NP_200311.1 3 - 4 236 86
NM_180862.1 NP_851193.1 3 - 4 231 86
GenBank mRNA AF174429.1 - 3 - 4 231 86
BT030093.1 - 2 - 3 150 86

View Full Product Details