Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIP2
Entrez Gene ID
835591
Gene Name
potassium channel tetramerization domain-containing protein / pentapeptide repeat-containing protein
Gene Aliases
AT5G55000, MBG8.27, MBG8_27
Chromosome Location
Chr.5: 22318305 - 22321805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124882.4 NP_200311.1 4 - 5 379 98
NM_180862.1 NP_851193.1 4 - 5 374 98
GenBank mRNA AF174429.1 - 4 - 5 374 98
BT030093.1 - 3 - 4 293 98

View Full Product Details