Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACA8
Entrez Gene ID
835815
Gene Name
autoinhibited Ca2+ -ATPase
Gene Aliases
''AUTOINHIBITED CA2+ -ATPASE, ''autoinhibited Ca2+ -ATPase, 'autoinhibited Ca2+ -ATPase, AT-ACA8, AT5G57110, AUTOINHIBITED CA2+ -ATPASE, ISOFORM 8, ISOFORM 8'', MUL3.5, MUL3_5, autoinhibited Ca2+ -ATPase, isoform 8, isoform 8', isoform 8''
Chromosome Location
Chr.5: 23109438 - 23117686 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345242.1 NP_001331281.1 4 - 5 521 65
NM_125093.6 NP_200521.3 4 - 5 570 65
NM_180869.3 NP_851200.1 4 - 5 412 65
GenBank mRNA AJ249352.1 - 4 - 5 414 65
AK226961.1 - 3 - 4 336 65
AY069869.1 - 4 - 5 414 65

View Full Product Details