Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APO2
Entrez Gene ID
835904
Gene Name
APO RNA-binding protein (DUF794)
Gene Aliases
ACCUMULATION OF PHOTOSYSTEM ONE 2, AT5G57930, MTI20.19, MTI20_19, emb1629, embryo defective 1629
Chromosome Location
Chr.5: 23454592 - 23456526 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125178.6 NP_200601.3 3 - 4 599 70
NM_180877.2 NP_851208.1 3 - 4 578 70
GenBank mRNA AX765838.1 - 3 - 4 501 70
AY062701.1 - 3 - 4 578 70
AY114672.1 - 3 - 4 492 70

View Full Product Details