Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHOT2
Entrez Gene ID
835926
Gene Name
phototropin 2
Gene Aliases
AT5G58140, AtPHOT2, K21L19.6, K21L19_6, NON PHOTOTROPIC HYPOCOTYL 1-LIKE, NPL1, phototropin 2
Chromosome Location
Chr.5: 23523825 - 23533111 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345288.1 NP_001318824.1 3 - 4 1163 68
NM_001345289.1 NP_001330355.1 2 - 3 802 68
NM_001345290.1 NP_001330356.1 1 - 2 1152 68
NM_125199.2 NP_568874.2 2 - 3 622 68
NM_180879.2 NP_851210.1 2 - 3 979 68
NM_180880.2 NP_851211.1 2 - 3 975 68
NM_180881.3 NP_851212.2 1 - 2 760 68
GenBank mRNA AF053941.2 - 1 - 2 517 68

View Full Product Details