Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GUS1
Entrez Gene ID
836246
Gene Name
glucuronidase 1
Gene Aliases
AT5G61250, AtGUS1, MFB13.2, MFB13_2, glucuronidase 1
Chromosome Location
Chr.5: 24632154 - 24635312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345460.1 NP_001330163.1 5 - 6 917 149
NM_125518.5 NP_200933.2 6 - 7 976 149
NM_180907.3 NP_851238.1 6 - 7 978 149
GenBank mRNA AK317449.1 - 6 - 7 968 149
AY099736.1 - 6 - 7 899 149
BT002603.1 - 5 - 6 818 149

View Full Product Details