Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SC35
Entrez Gene ID
836541
Gene Name
serine/arginine-rich splicing factor-like protein, putative
Gene Aliases
ARABIDOPSIS THALIANA ORTHOLOG OF HUMAN SPLICING FACTOR SC35, AT5G64200, ATSC35, At-SC35, MSJ1.4, MSJ1_4, ortholog of human splicing factor SC35
Chromosome Location
Chr.5: 25681202 - 25683641 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125816.4 NP_201225.1 7 - 8 994 102
NM_180930.3 NP_851261.1 7 - 8 994 102
GenBank mRNA AY063890.1 - 7 - 8 965 102
AY127004.1 - 7 - 8 983 102

View Full Product Details