Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASN2
Entrez Gene ID
836625
Gene Name
asparagine synthetase 2
Gene Aliases
ASPARAGINE SYNTHETASE 2, AT5G65010, MXK3.25, MXK3_25, asparagine synthetase 2
Chromosome Location
Chr.5: 25969035 - 25972575 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125900.2 NP_201306.2 2 - 3 252 95
NM_180941.3 NP_851272.1 2 - 3 407 95
GenBank mRNA AF367340.1 - 2 - 3 251 95
AY124866.1 - 2 - 3 218 95

View Full Product Details