Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGA1
Entrez Gene ID
836646
Gene Name
bZIP transcription factor family protein
Gene Aliases
AT5G65210, MQN23.15, MQN23_15, TGACG sequence-specific binding protein 1
Chromosome Location
Chr.5: 26057533 - 26061157 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037070.1 NP_001032147.1 5 - 6 610 127
NM_001037071.1 NP_001032148.1 5 - 6 517 127
NM_001037072.2 NP_001032149.1 6 - 7 537 127
NM_001085331.1 NP_001078800.1 5 - 6 826 127
NM_001345675.1 NP_001318881.1 5 - 6 847 127
NM_125919.3 NP_201324.1 4 - 5 1307 127
NM_180942.3 NP_851273.1 5 - 6 920 127
GenBank mRNA AK317142.1 - 5 - 6 518 127
AK317488.1 - 6 - 7 538 127
AY099593.1 - 5 - 6 636 127
AY128838.1 - 4 - 5 388 127

View Full Product Details