Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSL4
Entrez Gene ID
824305
Gene Name
strictosidine synthase-like 4
Gene Aliases
AT3G51420, ATSSL4, STRICTOSIDINE SYNTHASE-LIKE 4, strictosidine synthase-like 4
Chromosome Location
Chr.3: 19083980 - 19086300 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115001.3 NP_190710.1 2 - 3 812 83
GenBank mRNA AY087809.1 - 2 - 3 757 83
BT024894.1 - 2 - 3 710 83

View Full Product Details