Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMK2
Entrez Gene ID
824337
Gene Name
inflorescence meristem receptor-like kinase 2
Gene Aliases
AT3G51740, inflorescence meristem receptor-like kinase 2
Chromosome Location
Chr.3: 19188844 - 19192161 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115033.4 NP_190742.1 1 - 2 2278 124
GenBank mRNA AF372978.1 - 1 - 2 2039 124
AY054515.1 - 1 - 2 2073 124
BT006350.1 - 1 - 2 1874 124
FJ708739.1 - 1 - 2 1874 124

View Full Product Details