Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETO1
Entrez Gene ID
824340
Gene Name
tetratricopeptide repeat (TPR)-containing protein
Gene Aliases
ARABIDOPSIS ETHYLENE OVERPRODUCER 1, AT3G51770, ATEOL1, ETHYLENE OVERPRODUCER 1, ETHYLENE OVERPRODUCER1
Chromosome Location
Chr.3: 19199750 - 19204464 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035762.6 NP_001030839.5 2 - 3 2426 74
NM_115036.7 NP_190745.6 2 - 3 2420 74

View Full Product Details